missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dodecyl Sulfate, Ultrapure Bioreagent, J.T. Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

396.92 NOK - 43600.00 NOK

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym SDS
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15518244
View Documents
Avantor J.T.Baker
4095-00
25 g POLYSTORMOR This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15538244
View Documents
Avantor J.T.Baker
4095-04
100 g Poly Bottle
4125.00 NOK
Each
Please to purchase this item. Need a web account? Register with us today!
15533674
View Documents
Avantor J.T.Baker
4095-02
1 kg Poly bottle
25145.00 NOK
1kg
Please to purchase this item. Need a web account? Register with us today!
15553351
View Documents
Avantor J.T.Baker
4095-05
2 kg Poly Pail
43600.00 NOK
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

    For Electrophoresis and Protein Solubilization; For Laboratory,Research,or Manufacturing Use
Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Dodecyl Sulfate, Ultrapure Bioreagent, J.T. Baker™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.