missing translation for 'onlineSavingsMsg'
Learn More
Sodium dichromate dihydrate, ExpertQ, Scharlab Product Code.: 11358494

Sodium dichromate dihydrate, ExpertQ, Scharlab

Product Code. 11358494
500 g
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11358494

Brand: Scharlab SO02600500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Specifications

CAS 7789-12-0
Quantity 500 g
Molecular Formula Cr2H4Na2O9
InChI Key JYDRNIYTFCBIFC-UHFFFAOYSA-N
SMILES O.O.[Na+].[Na+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
IUPAC Name disodium [(oxidodioxochromio)oxy]chromiumoylolate dihydrate
Molecular Weight (g/mol) 298.00
Grade ExpertQ
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium dichromate dihydrate, ExpertQ, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.