missing translation for 'onlineSavingsMsg'
Learn More
Sodium carbonate, 99+%, for analysis, granular Product Code.: 10666422

Sodium carbonate, 99+%, for analysis, granular

Product Code. p-3737354 Shop All Thermo Scientific Chemicals Products
1 kg, Plastic Bottle, 1kg
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
Packaging:
Plastic Bottle
Plastic Drum
Unit Size:
1kg
2.50kg
5kg
This item is not returnable. View return policy

Product Code. 10666422

Brand: Thermo Scientific Acros 206800010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 497-19-8 | CNa2O3 | 105.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay sodaShow MoreShow Less
PubChem CID 10340
ChEBI CHEBI:29377
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Boiling Point 1600.0°C
Melting Point 851.0°C
Color White
Physical Form Fine Granular Powder
Quantity 1 kg
Assay Percent Range 99+%
Linear Formula Na2CO3
Fieser 7332
Merck Index 15, 8731
Solubility Information Solubility in water: 22 g/100mL (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol
IUPAC Name disodium;carbonate
Formula Weight 105.99
Percent Purity ≥99%
Grade Analytical
Loss on Drying 0.5% max. (285°C)
Packaging Plastic Bottle
Chemical Name or Material Sodium carbonate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0466923 Certificate TypeCertificate of Analysis Date30/10/2024 Product Code206800025,206800050,206800010
Lot NumberA0462639 Certificate TypeCertificate of Analysis Date16/05/2024 Product Code206800025,206800050,206800010
Lot NumberA0435032 Certificate TypeCertificate of Analysis Date15/04/2024 Product Code206800025,206800050,206800010
Lot NumberA0395371 Certificate TypeCertificate of Analysis Date14/12/2022 Product Code206800025,206800050,206800010
Lot NumberA0398841 Certificate TypeCertificate of Analysis Date14/12/2022 Product Code206800025,206800050,206800010
compliance-icons
Product Identifier
  • Sodium carbonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium carbonate, 99+%, for analysis, granular >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.