missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Caproate, MP Biomedicals
Click to view available options
Quantity:
100 g
Specifications
Specifications
CAS | 10051-44-2 |
Molecular Formula | C6H11NaO2 |
MDL Number | MFCD00059056 |
Quantity | 100 g |
Synonym | sodium hexanoate, sodium caproate, hexanoic acid, sodium salt, hexanoic acid sodium salt, sodium capronate, sodium n-caproate, sodium n-hexanoate, unii-e00cg11s66, caproic acid sodium salt, hexanoic acid, sodium salt 1:1 |
InChI Key | UDWXLZLRRVQONG-UHFFFAOYSA-M |
SMILES | [Na+].CCCCCC([O-])=O |
IUPAC Name | sodium hexanoate |
Molecular Weight (g/mol) | 138.14 |
PubChem CID | 4087444 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'