missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Cacodylate Trihydrate, 97+%, Extra Pure, SLR, Fisher Chemical
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Specifications
Specifications
Melting Point | 60°C |
CAS | 124-65-2 |
Color | White |
Physical Form | Solid |
Quantity | 100 g |
Molecular Formula | C2H6AsNaO2 |
MDL Number | 149079 |
Synonym | sodium dimethylarsinate, sodium cacodylate, alkarsodyl, cacodylic acid sodium salt, dimethylarsinic acid sodium salt, arsecodile, arsicodile, arsycodile, arsysodila, chemaid |
InChI Key | IHQKEDIOMGYHEB-UHFFFAOYSA-M |
SMILES | C[As](=O)(C)[O-].[Na+] |
Show More |
Safety and Handling



Product Identifier
- Sodium cacodylate trihydrate
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1A
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H350-May cause cancer.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Cacodylate Trihydrate, 97+%, Extra Pure, SLR, Fisher Chemical >
Spot an opportunity for improvement?Share a Content Correction