missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Borohydride, 98-99%, (Crystalline), MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
50 g
100 g
500 g
Specifications
Specifications
Physical Form | Crystalline |
Quantity | 25 g |
Assay Percent Range | 98-99% |
Formula Weight | 37.8 |
Chemical Name or Material | Sodium Borohydride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Borohydride, 98-99%, (Crystalline), MP Biomedicals >
Spot an opportunity for improvement?Share a Content Correction