Learn More
Thermo Scientific Chemicals Sodium bicarbonate, 1M buffer soln., pH 9.0
CAS: 144-55-8 | CHNaO3 | 84.01 g/mol
1080.00 NOK - 1580.00 NOK
Chemical Identifiers
CAS | 144-55-8 |
---|---|
Molecular Formula | CHNaO3 |
Molecular Weight (g/mol) | 84.01 |
MDL Number | MFCD00003528 |
InChI Key | UIIMBOGNXHQVGW-UHFFFAOYSA-M |
PubChem CID | 516892 |
ChEBI | CHEBI:32139 |
IUPAC Name | sodium;hydrogen carbonate |
SMILES | [Na+].OC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15442909
|
Thermo Scientific Chemicals
J60066.AK |
250 mL |
1080.00 NOK
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15452909
|
Thermo Scientific Chemicals
J60066.AP |
500 mL |
1580.00 NOK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
144-55-8 | |
84.01 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
CHEBI:32139 | |
[Na+].OC([O-])=O |
CHNaO3 | |
MFCD00003528 | |
516892 | |
sodium;hydrogen carbonate |
Specifications
144-55-8 | |
9.0 | |
CHNaO3 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
sodium;hydrogen carbonate | |
516892 | |
Sodium bicarbonate buffer soln. |
1 M | |
Liquid | |
MFCD00003528 | |
[Na+].OC([O-])=O | |
84.01 | |
CHEBI:32139 |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.