missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Acetate Trihydrate, ACS Reagent Grade, Scharlab
Click to view available options
Quantity:
1 kg
Description
- Synonyms: Acetic acid sodium salt trihydrate
- Molecular Formula: CH3COONa • 3H2O
- Molar Mass: 136.08g/mol
- CAS: 6131-90-4
- EIN ECS No.: 204-823-8
Specifications
Specifications
Boiling Point | >400°C (anhyd, decomposition) |
Melting Point | 58°C |
CAS | 6131-90-4 |
pH | 7.5–9.2 (5%, water) |
Quantity | 1 kg |
Molecular Formula | C2H9NaO5 |
Flash Point | >250°C (anhyd) |
Synonym | sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn |
InChI Key | AYRVGWHSXIMRAB-UHFFFAOYSA-M |
SMILES | CC(=O)[O-].O.O.O.[Na+] |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Acetate Trihydrate, ACS Reagent Grade, Scharlab >
Spot an opportunity for improvement?Share a Content Correction