missing translation for 'onlineSavingsMsg'
Learn More
Sodium Acetate Anhydrous, ACS Reagent Grade, Scharlab Product Code.: 11318484

Sodium Acetate Anhydrous, ACS Reagent Grade, Scharlab

Product Code. 11318484
1 kg, 1kg
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
This item is not returnable. View return policy

Product Code. 11318484

Brand: Scharlab SO00351000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in the food industry, analytical chemistry, the synthesis of organic products, and photography.

  • Synonyms: Acetic acid sodium salt anhydrous
  • Molecular Formula: CH3COONa
  • Molar Mass: 82.03g/mol
  • CAS: 127-09-3
  • EIN ECS No.: 204-823-8

Specifications

Boiling Point >400°C (decomposition)
Melting Point 324°C (decomposition)
CAS 127-09-3
pH 7.0–9.2 (5%, water) 
Quantity 1 kg
Molecular Formula C2H3NaO2
MDL Number MFCD00012459
Flash Point >250°C
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
SMILES [Na+].CC([O-])=O
IUPAC Name sodium acetate
Molecular Weight (g/mol) 82.03
PubChem CID 517045
ChEBI CHEBI:32954
Formula Weight 82.03g/mol
Percent Purity 99.0% min (titration with HClO4)
Grade ACS Reagent
Identification Passes Test
Ignition Point 607°C
Loss on Drying 1.0% max (120°C)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Acetate Anhydrous, ACS Reagent Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.