missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate, anhydrous, >98%, Molecular biology reagent grade, MP Biomedicals™

Product Code. 11466481
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 127-09-3
Color White
Physical Form Granular Powder
Quantity 1 kg
Molecular Formula C2H3NaO2
MDL Number MFCD00012459
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
SMILES [Na+].CC([O-])=O
IUPAC Name sodium acetate
Molecular Weight (g/mol) 82.03
PubChem CID 517045
ChEBI CHEBI:32954
Formula Weight 82.03
Percent Purity >98%
Grade Molecular Biology
Density 1.45g/cm3
Chemical Name or Material Sodium acetate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'