Learn More
Sodium 4-aminohippurate hydrate, 98%, Thermo Scientific Chemicals
Product Code. 11432438
Click to view available options
Quantity:
5 g
25 g
100 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 206658-83-5
Molecular Formula C9H10N2NaO3+
Molecular Weight (g/mol) 217.18
MDL Number MFCD00150723
InChI Key UNZMYCAEMNVPHX-UHFFFAOYSA-N
Synonym aminohippurate sodium
PubChem CID 57465078
IUPAC Name sodium;2-[(4-aminobenzoyl)amino]acetic acid
SMILES C1=CC(=CC=C1C(=O)NCC(=O)O)N.[Na+]
Specifications
Melting Point 123°C to 127°C
Quantity 25 g
Formula Weight 216.17 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Sodium 4-aminohippurate hydrate