Learn More
BRAND™ Soda-Lime Glass Watch Glasses
Stress-reduced to minimize risk of shattering.
Brand:  BRAND™ 150030
368.50 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 10558401
  • 468.00 NOK / Pack of 10
Elevate Your Beverage Testing with Precision Solutions
Use promo code 24135 at checkout
View Available Promotions
Only null left
Estimated Shipment: 15-07-2025
Description
  • Soda-lime glass to DIN 12341
  • Ground edges
  • Stress-reduced to minimize risk of shattering
TRUSTED_SUSTAINABILITY
Specifications
Specifications
70 mm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BRAND™ Soda-Lime Glass Watch Glasses > Diameter (Metric): 70 mm