Learn More
BRAND™ Soda-Lime Glass Watch Glasses
Stress-reduced to minimize risk of shattering.
Brand: BRAND™ 150040
374.35 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
- Soda-lime glass to DIN 12341
- Ground edges
- Stress-reduced to minimize risk of shattering
Specifications
DIN 12341 | |
Soda Lime Glass | |
Watch Glass | |
80 mm |
Flat | |
Round | |
Polished |
Product Content Correction
Product Title
BRAND™ Soda-Lime Glass Watch Glasses > Diameter: 80mm