Learn More
DWK Life Sciences Kimble Chase™ Soda Lime Glass Tube
Conduct research and experimentation with Soda Lime Glass Tube. This basic test tube has an open end, round bottom and a cap.
Brand: DWK Life Sciences F40752800D0W2
1068.75 NOK valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
- Reusable
- 200Pack
- Clear soda lime glass
- With cap
Specifications
75 mm | |
200Pack | |
28 mm | |
Test Tube |