Learn More
DWK Life Sciences Kimble Chase™ Soda Lime Glass Culture Tube
Use the Soda Lime Glass Culture Tube to view cell cultures in the laboratory. This disposable soda lime glass rim top tube is plain (unlabeled) and features a sturdy, uniform bottom.
Brand:  DWK Life Sciences A407011000811
102.60 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 11842383
  • 108.00 NOK / Pack of 200
Estimated Shipment: 30-04-2025
to see stock.
Explore more special offers
Description
  • Rimmed top
  • Unlableled
  • Clear soda lime glass
  • Uniform bottom
  • Disposable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
70 mm
200Pack
11 mm
Test Tube
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    24
    Hours
    15
    Minutes
    39
    Seconds
    15
Product Content Correction
Product Title
DWK Life Sciences Kimble Chase™ Soda Lime Glass Culture Tube > Diameter: 11mm; Length: 70mm