Learn More
BRAND™ Wire Handle for Staining Trough
Stainless steel construction. For use with staining trough. Order staining trough, tray and wire handle separately.
Brand: BRAND™ 473100
479.65 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Stainless steel construction
- For use with Staining Trough Tray
- Supplied in packs of 10
Specifications
Stainless Steel | |
Staining Trough Tray | |
Wire handle |
Product Content Correction
Product Title
BRAND™ Wire Handle for Staining Trough > Max. Slide Size: 76 x 26mm