Learn More
BRAND™ Soda Lime Glass Petri Dishes
Perfect for microbial or cell culture applications, these Soda Lime Glass Petri Dishes feature excellent glass quality and workmanship.
Brand: BRAND™ 455751
773.79 NOK valid until 2025-09-30
Use promo code "24345" to get your promotional price.
This item is not returnable.
View return policy
Description
- Dish and lid are flat inside and outside
- Free from bubbles and streaks
- Cut edges are fire-polished
- Suitable for short-term chemical exposure
Specifications
Soda Lime Glass | |
Petri Dish | |
25 mm |
Round | |
Fire-polished cut edges | |
150 mm |
Product Content Correction
Product Title
BRAND™ Soda Lime Glass Petri Dishes > Diameter: 150mm