Learn More
Brand™ Soda-lime Glass Oxygen Flasks
Brand: Brand™ 386038
652.79 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
100 to 150 mL | |
Soda Lime Glass |
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Product Content Correction
Product Title
Brand™ Soda-lime Glass Oxygen Flasks > Capacity: 100 to 150mL