missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX8 Antibody (OTI1A1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-45896
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunofluorescence
SNX8 Monoclonal specifically detects SNX8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.
Specifications
SNX8 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:150, Immunofluorescence 1:100 | |
DKFZp761E1721, Mvp1, sorting nexin 8, sorting nexin-8 | |
Mouse | |
52.4 kDa | |
0.1 mL | |
Signal Transduction | |
29886 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunofluorescence | |
OTI1A1 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
SNX8 | |
Full length human recombinant protein of human SNX8(NP_037453) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SNX8 Antibody (OTI1A1), Novus Biologicals™ > 0.1mL; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction