missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRPG Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15084-1-AP-150UL
This item is not returnable.
View return policy
Description
Core component of the spliceosomal U1, U2, U4 and U5 small nuclear ribonucleoproteins (snRNPs), the building blocks of the spliceosome. Thereby, plays an important role in the splicing of cellular pre-mRNAs. Most spliceosomal snRNPs contain a common set of Sm proteins SNRPB, SNRPD1, SNRPD2, SNRPD3, SNRPE, SNRPF and SNRPG that assemble in an heptameric protein ring on the Sm site of the small nuclear RNA to form the core snRNP. Appears to function in the U7 snRNP complex that is involved in histone 3′- end processing.Specifications
SNRPG | |
Polyclonal | |
Unconjugated | |
SNRPG | |
PBSCG, Sm G, Sm protein G, SMG, snRNP G, SNRPG | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6637, 68011 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P62308, P62309 | |
SNRPG | |
SNRPG Fusion Protein Ag7163 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction