Learn More
Motic™ SMZ-160-Trinocular head
This Trinocular Head is compatible with the SMZ-160 microscope.
Brand:  Motic™ 1101010100921
5285.07 NOK valid until 2025-12-20
Use promo code "24121" to get your promotional price.
Product Code. 17857285
  • ONLINE EXCLUSIVE PRICE
    6860.00 NOK
    6478.47 NOK / Each
    Save 381.53 NOK
    5% Off
View Available Promotions
Only null left
Estimated Shipment: 12-11-2025
Description
  • SMZ-160-Trinocular Head.
  • Includes 2 10x/20 mm eyepieces.
Specifications
Specifications
2 eyepieces of 10X/20 mm
None
Head only, need also a stand and light
SMZ-160 (Trinocular Head only)
None
10x/20 mm
10X
20 mm
5 years
Trinocular
Observation of Sample in 3D
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Motic™ SMZ-160-Trinocular head > Trinocular Head