Learn More
OHAUS™ Small Three Prong Sleeves
OHAUS™ Small Three Prong Sleeves are for use with clamps and can be easily removed for cleaning or replacement.
Brand: OHAUS™ 30400257
115.82 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Available in red or black vinyl and fiberglass
- Fiberglass sleeves are ideal for application above 100°C
Specifications
3 | |
Vinyl | |
Red |
Prong Sleeve | |
Small |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
OHAUS™ Small Three Prong Sleeves > Marerial: Vinyl; Color: Red