Learn More
OHAUS™ Small Three Prong Sleeves
OHAUS™ Small Three Prong Sleeves are for use with clamps and can be easily removed for cleaning or replacement.
Brand:  OHAUS™ 30400263
120.50 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15722679
  • 128.00 NOK / Each
Only null left
Estimated Shipment: 23-07-2025
Description
  • Available in red or black vinyl and fiberglass
  • Fiberglass sleeves are ideal for application above 100°C
TRUSTED_SUSTAINABILITY
Specifications
Specifications
3
Vinyl
Black
Prong Sleeve
Small
Certificates
Promotions
Promotions
Product Content Correction
Product Title
OHAUS™ Small Three Prong Sleeves > Marerial: Vinyl; Color: Black