Learn More
Small Sample Viscosity Standard ASTM D2162; UKAS ISO/IEC17025 and ISO 17034 certified, nominal kinematic viscosity 6684 cSt (40°C), Paragon Scientific™
Certified in strict accordance with ASTM D2162 at 40 °C and 100 °C under our ISO 17025 and ISO 17034 accreditation and manufactured specifically for the users of ASTM D7279, Standard Test Method for Kinematic Viscosity of Transparent and Opaque Liquids.
Brand: Paragon Scientific HVS14
This item is not returnable.
View return policy
Description
- Fully traceable to international standards
- Ensures full compliance to ASTM & IP test method protocol
- Certified in strict accordance with ASTM D2162
- 60mL lower volume cost effective solution
- Tamper evident security packaging
- 2 Year shelf life
- Manufactured in the United Kingdom
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Product Content Correction
Product Title
Small Sample Viscosity Standard ASTM D2162; UKAS ISO/IEC17025 and ISO 17034 certified, nominal kinematic viscosity 6684 cSt (40°C), Paragon Scientific™ > Quantity: 60 mL
Nominal values quoted. Certified Values will vary from batch to batch.