Learn More
SMAD7 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25840-1-AP-150UL
This item is not returnable.
View return policy
Description
The SMAD7 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human SMAD7. This antibody recognizes human, mouse, rat antigen. The SMAD7 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
SMAD7 | |
Polyclonal | |
Unconjugated | |
150 μL | |
4092 | |
IgG |
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA | |
540 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |