Learn More
Karl Hecht™ Assistent™ Sloping Stem Glass Funnels
For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Karl Hecht™ Assistent™ Sloping Stem Glass Funnels are manufactured from clear soda-lime glass.
Brand:  Karl Hecht™ 44000056
 View more versions of this product
Product Code. 13108365
  • 757.00 NOK / Each
Explore more special offers
Description
Description
  • Slope angle of 60°
  • Available in a range of diameters
Specifications
Specifications
200mm
Glass
60°
Laboratory Applications