Learn More
Karl Hecht™ Assistent™ Sloping Stem Glass Funnels
For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Karl Hecht™ Assistent™ Sloping Stem Glass Funnels are manufactured from clear soda-lime glass.
Brand:  Karl Hecht™ 44000036
 View more versions of this product
Product Code. 10093811
  • 70.00 NOK / Each
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Description
  • Slope angle of 60°
  • Available in a range of diameters
Specifications
Specifications
80mm
Glass
60°
Laboratory Applications