Learn More
Simport™ Scientific Slimsette™ Biopsy Processing/Embedding Cassette in Quickload™ stacks
Secure specimens during processing with Simport Scientific™ Slimsette™ Biopsy Processing/Embedding Cassette, in Quickload™ stacks.
Specifications
Material Acetal
Quantity 2000
View More Specs
Description
Description
In Quickload™ stacks
  • 45° Anterior writing area
  • Recessed cover
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
Specifications
Acetal
2000
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Simport™ Scientific Slimsette™ Biopsy Processing/Embedding Cassette in Quickload™ stacks