Learn More
Thermo Scientific™ Sliding Drawer Inserts
Maximize storage space with these sliding drawer inserts, for use with racks used in Thermo Scientific™ 13, 17, 23 and 28 cu. ft. ultra-low temperature freezers.
415.03 NOK
Specifications
Capacity (English) 28 cu. ft.
Capacity (Metric) 792.8 L
View More Specs
Products 1
Product Code Brand For Use With (Equipment) Description Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Description Price Quantity & Availability  
17134205
View Documents Promotion Details
Thermo Scientific™
1950666P5
13 cu. ft. (368L) capacity freezers Microplate insert
415.03 NOK
Pack of 5
Estimated Shipment: 11-03-2025
Description
Maximize storage space with Thermo Scientific™ Sliding Drawer Inserts, for use with racks used in Thermo Scientific™ 13, 17, 23 and 28 cu. ft. ultra-low temperature freezers.
  • Warranty varies by country, please contact us for details
Specifications
Specifications
28 cu. ft.
792.8 L