missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slanetz and Bartley Medium (Dehydrated)

Detect and enumerate enterococci from water and food samples with this selective medium.

2690.00 NOK

Specifications

Certifications/Compliance Industrial Reference: NMKL.
Form Powder
Product Type Dehydrated Culture Media
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10462643
View Documents Promotion Details
Thermo Scientific™
CM0377B
500 g
2690.00 NOK
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Detect and enumerate enterococci from water and food samples with Thermo Scientific™ Slanetz and Bartley Medium (Dehydrated). Slanetz and Bartley1 originally devised this medium to detect and enumerate enterococci by the technique of membrane filtration, but it has also proved useful as a direct plating medium2, 3. The medium is very selective for enterococci and, when it is incubated at elevated temperatures (44-45°C), all red or maroon colonies may be accepted as presumptive enterococci4, 5.

The detection and enumeration of enterococci is of primary importance for food hygiene monitoring and microbial quality of water and food.

The Environment Agency 'Microbiology of Drinking Water 2002'6 recommend the use of Slanetz and Bartley medium for the enumeration of enterococci in water supplies, as does ISO in the standard for water quality7. Food samples can be examined for enterococci by the method suggested by the Nordic Committee of Food Analysis3.

  • Easy-to-read: due to the reduction of tetrazolium chloride in the medium the enterococci form dark red/ maroon colonies.
  • Selective: sodium azide in the medium is selective against Gram-negative organisms.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

Specifications

Specifications

Industrial Reference: NMKL.
Powder
Dehydrated Culture Media
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • SLANETZ and BARTLEY MEDIUM

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : Sodium azide

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Slanetz and Bartley Medium (Dehydrated)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.