Learn More
RETSCH Sintered Alumina I Grinding Balls
Brand:  RETSCH 05.368.0021
Product Code. 10593451
  • 2225.00 NOK / Each
Only null left
Estimated Shipment: 26-08-2025
Specifications
Grinding Balls
0.9 mm
MM 200 and MM 301
Sintered Alumina I
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
RETSCH Sintered Alumina I Grinding Balls > Dia.: 0.9mm