Learn More
Vitlab™ Simplex II Fix Dispenser
Vitlab™ Simplex II Fix Dispenser is designed to be practically universally applicable and can be used with many organic and inorganic solutions.
Brand: Vitlab™ 1622504
This item is not returnable.
View return policy
Description
- Bottle-top dispenser with fixed volume
- Direct displacement plunger with broad PFA seal prevents crystallisation build-up
- Screwable ventilation plug for easy connection of a drying tube
- Screwable discharge valve is equipped with an additional safety bulb and closes the dosing channel, if dispensing tube is not installed, so that no medium can escape
- Equipped with a positive displacement piston and a fluoroplastic (PFA) sealing lip on the cylinder wall
- Nominal volume 1 to 10mL includes adapters GL 25, GL 28, GL 32, GL 38, S 40 and telescopic intake tube (length 125 to 240mm)
Specifications
5 mL |
Product Content Correction
Product Title
Vitlab™ Simplex II Fix Dispenser > Volume: 5 mL