Learn More
Vitlab™ Simplex II Bottletop Dispenser
Vitlab™ Simplex II Bottletop Dispenser is designed to be practically universally applicable and can be used with many organic and inorganic solutions.
Brand:  Vitlab™ 1621504
2669.93 NOK valid until 2025-06-30
Use promo code "24075" to get your promotional price.
 View more versions of this product
Product Code. 15624854
  • 4010.00 NOK / Each
The Ultimate Lab Appliances Promotion
Use promo code 24075 at checkout
View Available Promotions
Only null left
Estimated Shipment: 14-07-2025
Description
  • Direct displacement plunger with broad PFA seal prevents crystallisation build-up
  • Settings are quickly and accurately adjusted due to interior scalloped track and practical screw mechanism
  • Screwable ventilation plug for easy connection of a drying tube
  • Screwable discharge valve is equipped with an additional safety bulb and closes the dosing channel, if dispensing tube is not installed, so that no medium can escape
  • Equipped with a positive displacement piston and a fluoroplastic (PFA) sealing lip on the cylinder wall
  • Nominal volume 2 to 10mL includes adapters GL 25, GL 28, GL 32, GL 38, S 40 and telescopic intake tube (length 125 to 240mm)
  • Nominal volume 25 to 100mL includes adapters GL 32, GL 38, S 40 and telescopic intake tube (length 170 to 330mm)
Specifications
Specifications
0.5 to 5.0 mL
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.
    Promo Code: 24075
    Expires2025-06-30
Product Content Correction
Product Title
Vitlab™ Simplex II Bottletop Dispenser > Volume: 0.5 to 5.0 mL