missing translation for 'onlineSavingsMsg'
Learn More

Silver solution for ICP 10g/L HNO3 matrix, Fisher chemical™
BIOPHARMA

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

1220.00 NOK - 4110.00 NOK

Specifications

Analyte or Component Names Ag
Concentration 10±0.02 g/L
Type Mass Spectrometry Standard
Element Ag
Solvent or Matrix HNO3

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
12936244
BIOPHARMA
View Documents
Fisher Chemical
S 201 100
100 mL Bottle
1220.00 NOK
100mL
Please to purchase this item. Need a web account? Register with us today!
12946244
View Documents
Fisher Chemical
S 201 250
250 mL Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12956244
View Documents
Fisher Chemical
S 201 500
500 mL Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

Ag
Mass Spectrometry Standard
HNO3
10±0.02 g/L
Ag
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silver solution for ICP 10g/L HNO3 matrix, Fisher chemical™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.