Learn More
Silver gauze, 80 mesh woven from 0.115mm (0.0045in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11325247
Silver gauze, 80 mesh woven from 0.115mm (0.0045in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals
Product Code. 11325247
75 x 75 mm
Click to view available options
Quantity:
25 x 25 mm
75 x 75 mm
150 x 150 mm
300 x 300 mm
Product Code. 11325247
Brand: Thermo Scientific Alfa Aesar 040936.FL
Request Bulk or Custom Format
Silver gauze is used for filtration. It is also used in electrodes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]
Specifications
Color Silver
Appearance Silver-colored guaze
Assay Percent Range 99.9% (Metals basis)
Quantity 75 x 75 mm
Solubility Information Insoluble in water.
Mesh Size 80 Mesh
Odor Characteristic
Form Wire Cloth
Chemical Name or Material Silver
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ12I051 Certificate TypeCertificate of Analysis Date22/04/2022 Catalog Number040936.HB,040936.GT,040936.FF,040936.FL
Lot NumberR19G015 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number040936.HB,040936.GT,040936.FF,040936.FL
Lot NumberR23E008 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040936.HB,040936.GT,040936.FF,040936.FL
Lot NumberI13S022 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040936.HB,040936.GT,040936.FF,040936.FL
Lot NumberX22H008 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040936.HB,040936.GT,040936.FF,040936.FL
Product Title
Silver gauze, 80 mesh woven from 0.115mm (0.0045in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals >