Learn More
Silver Conductive Ink Product Code.: 15402437
Silver Conductive Ink
Product Code. 15402437
25 g, 25g
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 15402437
Brand: Thermo Scientific Alfa Aesar 045661.14
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Silver Conductive Ink
Density 2.5
MDL Number MFCD01868501
Quantity 25 g
Form Liquid
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberQ11L029 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code045661.14,045661.06,045661.22
Lot NumberZ12K010 Certificate TypeCertificate of Analysis Date23/01/2025 Product Code045661.14,045661.06,045661.22
Lot NumberT01K046 Certificate TypeCertificate of Analysis Date23/07/2024 Product Code045661.14,045661.06,045661.22
Lot NumberW12I003 Certificate TypeCertificate of Analysis Date16/09/2022 Product Code045661.14,045661.06,045661.22
Lot NumberT06I077 Certificate TypeCertificate of Analysis Date19/07/2022 Product Code045661.14,045661.06,045661.22
compliance-icons compliance-icons
Product Identifier
  • Silver Conductive Ink
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Silver, Diethylene glycol monoethyl ether acetate
Product Title
Silver Conductive Ink >