Learn More
Thermo Scientific Chemicals Silver Conductive Ink
Brand:  Thermo Scientific Chemicals 044958.22
 View more versions of this product
Product Code. 15492077
  • 13460.00 NOK / 100g
Description
This is a conducting screen printable thick film ink for membrane switch production.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Conductive Ink
100 g
Silver
Membrane switch production
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU15J065 Certificate TypeCertificate of Analysis Date06/09/2023 Catalog Number044958.06,044958.14,044958.22
Lot NumberR05I037 Certificate TypeCertificate of Analysis Date21/05/2022 Catalog Number044972.03,044958.06,044972.06,044958.14,044958.22
Lot NumberP08I014 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number044972.03,044958.06,044972.06,044958.14,044958.22
Lot NumberM24E001 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044972.03,044958.06,044972.06,044958.14,044958.22
Lot NumberY22F060 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044972.03,044958.06,044972.06,044958.14,044958.22
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific Chemicals Silver Conductive Ink > 100g