Learn More
BINDER SILVER 3-year Maintenance Contract
Maintenance service as contractually agreed.
7097.03 NOK - 15935.40 NOK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
30274266
|
BINDER
DL20-0825 |
BINDER Chambers Series KBF-S, KBF-S ECO |
11150.39 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30274292
|
BINDER
DL20-0855 |
BINDER Chambers Series UF V |
7097.03 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30274308
|
BINDER
DL20-0875 |
BINDER Chambers Series CB, CB-S |
12321.04 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30274285
|
BINDER
DL20-0860 |
BINDER Chambers Series VDL |
15935.40 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30274277
|
BINDER
DL20-0845 |
BINDER Chambers Series KT, KB, KB ECO |
7097.03 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30274276
|
BINDER
DL20-0815 |
BINDER Series B, BD, BD-S, E, ED, ED-S |
7097.03 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30274270
|
BINDER
DL20-0835 |
BINDER Chambers Series MKF, MKFT |
12745.40 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30274267
|
BINDER
DL20-0865 |
BINDER Chambers Series CBF |
13184.39 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30274304
|
BINDER
DL20-0880 |
BINDER Chambers Series VD |
13184.39 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Maintenance service as contractually agreed
- Visual inspection of mechanical and electrical components
- Check of control response
- 20% discount on spare parts
- Testing of all key functions
- Calibration of one test temperature in the center of the usable space as specified by the user
- Without certificate
Specifications
Maintenance Service |
Product Title
BINDER SILVER 3-year Maintenance Contract