Learn More
Uvex Silv-Air Classic 2312 FFP3 Mask
Shop All Uvex Products
Click to view available options
:
Filter Type: FFP3
Description
- With headband loops and integrated sealing lips in all protection classes, it is really easy to fit the masks correctly.
- The masks can also be worn comfortably for a longer period of time without becoming bothersome.
Specifications
Specifications
Aerosol Type | P - Oil-Proof |
Color | White |
Design Type | Cup/Molded |
Disposable | Reusable |
Filter Type | FFP3 |
Material | Polypropylene, Polyester |
Product Type | Healthcare Particulate Respirator and Surgical Mask |
Size | Medium |
With Exhalation Valve | Yes |
Latex Free | Yes |
Show More |
Product Content Correction
Product Title
Uvex Silv-Air Classic 2312 FFP3 Mask >