Learn More
Thermo Scientific™ SilTite™ GC Capillary Column Connector
Use SilTite GC Capillary Column Connectors for fused silica capillary GC columns.
Brand:  Thermo Scientific™ 290MU500
1312.49 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10301055
  • 1385.00 NOK / Each
Only null left
Estimated Shipment: 23-06-2025
Description
Use Thermo Scientific™ SilTite Capillary Column Connectors for fused silica capillary GC columns. They are glass lined for inertness and have a low thermal mass.
Join together two GC columns
Repair broken columns
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Capillary Column Connector
290 MU500
0.45 to 0.53 mm
Fused Silica Capillary Column
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ SilTite™ GC Capillary Column Connector > 1 Connector, 2 Nuts and 5 Ferrules (32mm I.D.)