Learn More
Thermo Scientific™ SilTite™ GC Capillary Column Connector
Use SilTite GC Capillary Column Connectors for fused silica capillary GC columns.
Brand:  Thermo Scientific™ 290MU499
1312.49 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10117864
  • 1385.00 NOK / Each
Only null left
Estimated Shipment: 19-06-2025
Description
Use Thermo Scientific™ SilTite Capillary Column Connectors for fused silica capillary GC columns. They are glass lined for inertness and have a low thermal mass.
Join together two GC columns
Repair broken columns
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Capillary Column Connector
290 MU499
32 mm
Fused Silica Capillary Column
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ SilTite™ GC Capillary Column Connector > 1 Connector, 2 Nuts and 5 Ferrules (0.1 - 0.25mm I.D.)