Learn More
Trajan Scientific and Medical™ SilTite™ Ferrules
Ensures easy, leak-free installation for capillary columns without the use of any tools.
Brand:  Trajan Scientific and Medical™ 073330
 View more versions of this product
Product Code. 15268923
  • 880.00 NOK / Pack of 10
Only null left
Estimated Shipment: 14-07-2025
Description
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Specifications
SilTite Ferrules for 0.1 to 0.25 mm ID Column for Thermo Scientific
For Use with Thermo GC/MS Interface Connections 0.10 to 0.25 mm I.D. Columns
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Trajan Scientific and Medical™ SilTite™ Ferrules > For use with Thermo GC/MS Interface Connections 0.10 to 0.25mm I.D. columns