Learn More
Thermo Scientific™ SilTite™ Ferrule Kits
Eliminate leaks with Thermo Scientific™ SilTite Ferrule Kits.
Brand:  Thermo Scientific™ 290MA217
2146.32 NOK valid until 2025-02-28
Use promo code "24209" to get your promotional price.
 View more versions of this product
Product Code. 10086154
  • 2259.28 NOK / Pack of 10
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Description
Eliminate leaks with Thermo Scientific™ SilTite Ferrule Kits. The ferrule forms a strong, permanent, airtight seal around the capillary column.
  • SilTite metal ferrules and nuts are made from the same material and have the same thermal expansion coefficient
  • Base of the ferrule is flat and forms a perfect seal with the MS interface
  • The ferrule's temperature tolerance is well above the limit of the injector, MS interface or GC oven
  • Unlike other ferrules, do not need re-tightening after installation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ferrule Kit
SilTite Kit for Thermo Scientific TRACE 1300/1310 GC SSL Injectors
Two SilTite Nuts and Ten Ferrules
290 MA217
SilTite Metal
0.53 mm