missing translation for 'onlineSavingsMsg'
Learn More

Silmitasertib, MedChemExpress

MedChemExpress Silmitasertib (CX-4945) is an orally bioavailable, highly selective and potent CK2 inhibitor, with IC50 values of 1 nM against CK2α and CK2α'.

Encompass_Preferred

1564.57 NOK - 10778.14 NOK

Specifications

Chemical Name or Material Silmitasertib
CAS 1009820-21-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H12ClN3O2
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16498108
Encompass_Preferred
View Documents
Medchem Express
HY-50855
10 mg
2544.40 NOK
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16480568
Encompass_Preferred
View Documents
Medchem Express
HY-50855
5 mg
1564.57 NOK
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16474027
Encompass_Preferred
View Documents
Medchem Express
HY-50855
50 mg
6858.82 NOK
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16447876
Encompass_Preferred
View Documents
Medchem Express
HY-50855
100 mg
10778.14 NOK
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Silmitasertib (CX-4945) is an orally bioavailable, highly selective and potent CK2 inhibitor, with IC50 values of 1 nM against CK2α and CK2α'.
Specifications

Specifications

Silmitasertib
Yellow
C19H12ClN3O2
DMSO : ≥ 35 mg/mL (100.07 mM) ∣0.1 M NaOH : 33.33 mg/mL (95.29 mM; ultrasonic and adjust pH to 9 with NaOH)
349.77
96.55%
Solid
1009820-21-6
Research
CX-4945
O=C(C1=CC=C2C3=C(C(NC4=CC=CC(Cl)=C4)=NC2=C1)C=CN=C3)O
349.77
Research
Cancer-Kinase/protease
Videos
Safety and Handling

Safety and Handling

H302∣H315∣H319∣H332∣H335

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Silmitasertib, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.