Learn More
Masterflex TUBING SILICONE PLAT LS 24, 7.6m
Brand:  Masterflex 96410-24
TUBING SILICONE PLAT LS 24, 7.6m
Product Code. 10183442
  • 2335.00 NOK / 7.60m
Only null left
Estimated Shipment: 03-09-2025
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Masterflex TUBING SILICONE PLAT LS 24, 7.6m >