Learn More
Fisherbrand™ Silicone Stoppers
For laboratory and/or food testing, industrial masking or educational use.
Brand:  Fisherbrand™ SB013/S
 View more versions of this product
Product Code. 10131511
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Non-toxic, translucent
  • Temperature range -60°C to 200°C
  • High electrical resistance
  • Resists cracking and spallation
  • High resistance to many chemicals including ozone
Specifications
Specifications
10/Pk.
13 mm
Silicone
Stopper
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Silicone Stoppers > Diameter: 13mm