Learn More
Fisherbrand™ General Purpose Silicone Sheet
An excellent choice for applications requiring good sealing properties at high and low temperatures. Fisherbrand™ General Purpose Silicone Sheets are also suitable for applications requiring direct food contact.
Brand:  Fisherbrand™ 1171-4365
 View more versions of this product
Product Code. 11714365
  • 6735.00 NOK / Pack of 5
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Description
  • Available plain or adhesive backed
  • Suitable for use at -40° to +200°C and +230°C short-term
  • Can be supplied in sheet form or converted into strips, seals, washers, gaskets and fabricated articles
Specifications
Specifications
1 x 360 x 360mm
Product Title
Fisherbrand™ General Purpose Silicone Sheet > Thickness: 1mm