missing translation for 'onlineSavingsMsg'
Learn More
Silicon(IV) oxide, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11303438

Silicon(IV) oxide, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11303438
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11303438

Brand: Thermo Scientific Alfa Aesar 036346.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Silica is used primarily in the production of glass for windows, drinking glasses, beverage bottles. The majority of optical fibers for telecommunication are also made from silica. It is a primary raw material for many ceramics such as earthenware, stoneware, and porcelain. The main ingredient in sand casting for the manufacture of a large number of metallic components in engineering and other applications. Silica is a common additive in the production of foods. An estimated 95% of silicon dioxide produced is consumed in the construction industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Boiling Point 2230°C
Melting Point 1710°C
Physical Form Fused Lump
Quantity 100 g
Assay Percent Range 99.99% (metals basis)
Merck Index 14,8493
Solubility Information Insoluble in water.
Formula Weight 60.09
Percent Purity 99.99%
Odor Odorless
Density 2.2 g/mL to 2.6 g/mL
Assay (metals basis)
Chemical Name or Material Silicon(IV) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP05K041 Certificate TypeCertificate of Analysis Date08/04/2024 Catalog Number036346.14,036346.22
Lot NumberZ12I015 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number036346.14,036346.22
Lot NumberW01G024 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number036346.14,036346.22
Lot NumberM17I028 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number036346.14,036346.22
Lot NumberR03H052 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number036346.14,036346.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silicon(IV) oxide, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.