Learn More
Thermo Scientific Chemicals Silica gel, C18-RP, 23%C, ca.1.2mmol/g, part. size 40-63μm
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatography
Specifications
Specifications
Packaging | Glass Bottle |
Quantity | 250 g |
Material | Silica Gel |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0464564 | Certificate of Analysis | 23/07/2024 | 360110500,360112500 |
A0462915 | Certificate of Analysis | 23/05/2024 | 360110500,360112500 |
A0458211 | Certificate of Analysis | 05/12/2023 | 360110500,360112500 |
A0455295 | Certificate of Analysis | 25/08/2023 | 360110500,360112500 |
A0452579 | Certificate of Analysis | 27/04/2023 | 360110500,360112500 |
Safety and Handling
- Silicagel, C18-Reversed Phase