Learn More
Silica Gel 60 Angstrom Pore Size, 60-200 Micron, for Column Chromatography, Fisher Chemical™
Brand: Fisher Chemical S/0694/60
834.47 NOK valid until 2025-08-24
Use promo code "24140" to get your promotional price.
This item is not returnable.
View return policy
Specifications
Plastic Powder Jar | |
1 kg |
Product Content Correction
Product Title
Silica Gel 60 Angstrom Pore Size, 60-200 Micron, for Column Chromatography, Fisher Chemical™ > 1kg, Plastic powder jar